Would you like to log in again?
MOTOR BLOWER ASSY POWERFLO
Supplier: 3M OCCUPATIONAL HEALTH
Motor Blower^ This assembly is a component of the 3M* Powerflow* System 240-01-00LBC. This lightweight unit (8.0 oz.) delivers a minimum of 4 cfm when used with the required high efficiency filter.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MOTOR BLOWER ASSY POWERFLO 67101-096 7000126344 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty