Would you like to log in again?
FIT TEST ADAPTER
Supplier: 3M OCCUPATIONAL HEALTH
Fit Test Adapter^ Use to conduct quantitative fit testing on 3M* Facepieces 6000 Series or 3M* Facepieces 7000 Series with bayonet style cartridge/filter holders.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FIT TEST ADAPTER 67101-100 7000001945 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty