Would you like to log in again?
POWERFLO SYSTEM FL FACE MD CS1
Supplier: 3M OCCUPATIONAL HEALTH
Powerflow* System^ This respirator is a lightweight, face-mounted Powered Air Purifying Respirator (PAPR). It uses a single high efficiency filter for protection against dusts, mists, fumes and radionuclides, making it ideal for lead, asbestos and other hazardous particulates.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
POWERFLO SYSTEM FL FACE MD CS1 67101-102 7000052156 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty