Would you like to log in again?
EYEGLASS FRAME AND MOUNT KIT
Supplier: 3M OCCUPATIONAL HEALTH
Eyeglass Frame and Mount Kit^ Use with 3M* Full Facepieces 7000 Series and wearer's corrective lenses. Do not use with 3M* Face Mounted PAPR Systems 43100-068 or 43100-068 , 3M* Welder's Faceshield 7988 or 3M* Welder's Faceshield Kit 7998.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EYEGLASS FRAME AND MOUNT KIT 67101-120 7000125980 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty