Would you like to log in again?
SPEAKING DIAPHRAGM KIT
Supplier: 3M OCCUPATIONAL HEALTH
Speaking Diaphragm Kit^ Replacement part for 3M* Full Facepieces 7000 Series. Includes speaking diaphragm, gasket, retainer and removal tool.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPEAKING DIAPHRAGM KIT 67101-122 7000051802 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty