Would you like to log in again?
AIR FILTER AND REGULAR PANEL
Supplier: 3M OCCUPATIONAL HEALTH
Air Filter And Regulator Panel^ This panel is designed to filter and regulate air being delivered to the supplied air respirator. It has a maximum capacity of 50 cfm at 105 psig. Note: This unit will NOT remove carbon monoxide (CO) from compressed air source.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
AIR FILTER AND REGULAR PANEL 67101-258 7000005415 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty