Would you like to log in again?
FILTER CARTRIDGE F/W-2806 CS10
Supplier: 3M OCCUPATIONAL HEALTH
Filter Cartridge^ This cartridge is 99.1% efficient against 0.4-0.6 micron particles.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER CARTRIDGE F/W-2806 CS10 67101-262 7000005377 Case of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty