Would you like to log in again?
COALESCING FILTER ASSY
Supplier: 3M OCCUPATIONAL HEALTH
Coalescing Filter Assay^ Used where the concentration of liquid aerosols in compressed air are greater than what can be filtered by the 3M* Compressed Air Filter and Regulating Panel W-2806 (67101-258) alone.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COALESCING FILTER ASSY 67101-288 7000005397 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty