Would you like to log in again?
MOTOR AND HOUSING ASSY. ULTRA
Supplier: 3M OCCUPATIONAL HEALTH
Motor And Housing Assembly^ Includes a high performance motor. Designed to generate more airflow to the wearer. Can be used with the 3M* Airstream* Headgear-Mounted Powered Air Purifying Respirator (PAPR) Systems 56221-876 and 56221-878. Intrinsically safe.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MOTOR AND HOUSING ASSY. ULTRA 67101-324 7000126510 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty