Would you like to log in again?
VISOR ANTI-FOG CS10
Supplier: 3M OCCUPATIONAL HEALTH
Anti-fog Visor^ Visor has anti-fog and scratch-resistant properties built-in. It can be used with the 3M* Airstream* Headgear-Mounted Powered Air Purifying Respirator (PAPR) System AS-200LBC (56221-874), AS400LBC (56221-876) and AS-600LBC (56221-878). Good solution for hot, humid environments.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VISOR ANTI-FOG CS10 67101-328 7100090721 Case of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty