Would you like to log in again?
INDL INTERCHGE SOCKET 3/8INMPT
Supplier: 3M OCCUPATIONAL HEALTH
Industrial Interchange Socket^ This socket is a standard fitting used with the 3M* Compressed Air Filter and Regulator Panel W-2806 (67101-258). It is a 3/8 inch MPT high pressure socket made of brass.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INDL INTERCHGE SOCKET 3/8INMPT 67101-336 7000005401 Case of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty