Would you like to log in again?
CONNECTOR LOW PRESSURE
Supplier: 3M OCCUPATIONAL HEALTH
Low Pressure Connector^ Used with the 3M* Supplied Air Hose W-3020. It is part of the 3M* Low Pressure Air Regulating Kit W-3197 which is used in continuous flow and pressure demand applications.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CONNECTOR LOW PRESSURE 67101-346 7010318282 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty