Would you like to log in again?
WAIST BELT VINYL
Supplier: 3M OCCUPATIONAL HEALTH
Vinyl Waist Belt^ This belt is part of the 3M* Continuous Flow, Pressure Demand and the Face-Mounted Powered Air Purifying Respirator (PAPR) Systems. It is adjustable to fit most sizes. Along with the belt clip assembly, it holds the supplied air hose in place. Vinyl material to resist contaminants.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WAIST BELT VINYL 67101-352 7000130997 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty