Would you like to log in again?
SHRADER PLUG 1/4 MPT CS2
Supplier: 3M OCCUPATIONAL HEALTH
Schrader Plug^ Used on the 3M* Connector W-3195 (67101-346), Low Pressure, which is used with the 3M* Supplied Air Hose W-3020, Low Pressure. It has a 1/4 inch MPT steel fitting.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHRADER PLUG 1/4 MPT CS2 67101-356 7010346759 Case of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty