Would you like to log in again?
Chemical Resistant Polycarbonate Face Shields, Oberon
Supplier: Oberon
128AFR 028 128R 2128R
10856-536PK 553.52 USD InStock
10856-536 10856-538 10856-378 67200-101
Chemical Resistant Polycarbonate Face Shields, Oberon
Face Shields
Lightweight and durable design protects users from potentially harmful chemicals.
  • Equipped with ratchet headgear
  • Coated by Silicote-6, to protect polycarbonate from many mild acids, bases, and solvents
Chemical resistant polycarbonate face shields are available with a variety of coatings. The replacement shield is designed for use with ratchet headgear and slot-fit adapters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty