Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dextrose 5% in water for injection, sterile

Supplier: BAXTER IV SOLUTIONS
Synonyms: Corn sugarD-GlucoseD-(+)-DextroseGlucolin(2R,3S,4R,5R)-2,3,4,5,6-PentahydroxyhexanalGlucoseDextrose

2B0081
68000-142CS 1715.71 USD InStock
68000-142
Dextrose 5% in water for injection, sterile
D(+)-Glucose
Sterile solutions for injection.

  • Intravenous use
  • Subcutaneous use
  • mOsmol/L (calc): 252
  • NDC Number: 0338-0017-11
  • pH: 4.0 (3.2–6.5)

Packaging: Multi Pack contains 16 bags per overwrap pouch. Quad Pack contains four bags per overwrap pouch. Single Pack contains one bag per overwrap pouch.


Formula: C₆H₁₂O₆
MW: 180.16 g/mol
MDL Number: MFCD00148912
CAS Number: 50-99-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty