Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Irrigation Set, TUR/Bladder Y-Type, Regulating Clamp, Baxter

Supplier: Baxter
2C4041
68000-730CS 770.02 USD InStock
68000-730
Irrigation Set, TUR/Bladder Y-Type, Regulating Clamp, Baxter
Infusions
For use with Baxter irrigating solutions in plastic containers such as UROMATIC or ARTHROMATIC plastic containers.

  • Contains DEHP
  • Sterile Package
  • Non pyrogenic

Approximate Length 82" (2.1 m)

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty