Would you like to log in again?
AQUASONIC® 100 Ultrasound Transmission Gel, Parker Lab Inc.
Supplier: Parker
01-02 01-34 01-08 01-50
68200-712CS 274.14 USD InStock
68200-712 68200-710 68200-706 68200-058
AQUASONIC® 100 Ultrasound Transmission Gel, Parker Lab Inc.
Ultrasound Gels
Widely used gel for diagnostic and therapeutic medical ultrasound which is completely aqueous, will not stain clothing or damage transducers.
  • Acoustically correct for the broad range of frequencies used
  • Unique "can't be copied" formula is bacteriostatic, non-sensitizing and non-irritating
  • Applicable for all procedures where a viscous gel is required
  • No formaldehyde
  • Not a spermicide
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty