Would you like to log in again?
Quiltec® I Wipers
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Quiltec® I
QT1-1212B/100 QT1-99 QT1-1212 LWQS0002 LWQT0019 QT1-44B LWQS0001 QT1-99B
75996-814CS 377.83 USD InStock
75996-814 89074-012 68300-560 68311-024 68310-222 68311-025 68310-218 68310-215
Quiltec® I Wipers
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Quiltec® I Wipes offer the highest sorptive capacity of any sealed edge wipe available. They are manufactured using two layers of knitted polyester fabric that are laminated together to form a stronger, more adsorbent wipe. Additional fluid is captured within the quilted pockets between the fabric layers, increasing the sorbent capacity. Quiltec® I have more than twice the adsorbent capacity of a single-layer wipe.
  • Extremely low in both particles and fibers, reducing possibility of contamination
  • Strong fabric is abrasion and chemical resistant
  • Excellent sorbency with solvents
These wipes have laser-cut, heat-sealed edges which reduce the number of releasable particles and fibers. They are laundered and packed in a class 4 cleanroom. Sterile Quiltec® wipes are sterilized via gamma irradiation, and a certificate of sterility is provided with each shipment.
Certifications: Recommended for ISO Class 3-5 environments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty