Would you like to log in again?
CONSTIX®, Sealed Foam Swabs, SF-13 Series, Contec®
Supplier: Contec
CONSTIX®
SF-13
68310-254CS 148.84 USD InStock
68310-254
CONSTIX®, Sealed Foam Swabs, SF-13 Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SF-13 swabs consist of 100ppi open-cell foam tightly sealed around the tip of a glass-filled polypropylene handle. The foam is secured to the handle without the use of adhesives or binders.
  • Polyurethane foam head provides good particle entrapment and removal
The foam head conforms tightly to the pointed shape of the swab handle to provide a very fine tip that enables access to small areas and crevices. The polyurethane foam head provides good particle entrapment and removal and is very low in particles and fibers.
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal. Though compatible with many common solvents, the SF-13 swab is not recommended for use with ketones such as acetone or MEK.
Ordering information: Laundered and double bagged.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty