Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PROSAT® Sterile Meltblown Polypropylene Wipes, Contec®

Supplier: Contec

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PROSAT®
PS-911EB/ETOH PS-911EB/ETOH
68310-355CS 1185.45 USD InStock
68310-355 CA68310-355
PROSAT® Sterile Meltblown Polypropylene Wipes, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
PROSAT® Sterile Polypropylene wipes are presaturated with 70% denatured ethanol and 30% deionized water. The meltblown polypropylene wipes provide a consistent release of solvent to thoroughly remove surface contaminants in critical environments. These wipes are exceptionally clean and also free from additives of any kind. This particular meltblown polypropylene contains very low levels of sodium and other ions.

  • Suitable for Grade A/B or ISO Class 5 environments
  • Validated sterile via gamma irradiation
  • Meltblown polypropylene fabric is exceptionally clean and free from additives
  • Low in particles and fibers
  • Ideal for product contact areas
  • Especially suitable for the metered release of 70% alcohol
  • Presaturated wipe reduces VOC emissions and solvent use
  • Resealable pouch preserves cleanliness and solvent saturation levels

PROSAT® Sterile Polypropylene wipes are validated sterile and compatible with Grade A/B or ISO Class 5 environments. PROSAT® meltblown polypropylene wipes are ideal for wiping articles prior to pass through, cleaning process tools and other equipment and general surface cleaning.

Presaturated wipes ensure consistent saturation of each wipe independent of operator. Presaturated wipes can increase solvent control and accountability as well as reduce VOC emissions. The wipes are provided in convenient and easy to use peel and reseal pouches.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty