Would you like to log in again?
SATWipes® Sigma™ Wipes, Contec®
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SATWipes®
SAT-C1-100 SAT-C1-7030 SAT-C1-7030 SWC30029 SWC30064 SAT-C1-100 SAT-C1-1090 SAT-C1-7030/18 SWC30029 SWC30064 SAT-C1-1090 SAT-C1-7030/18
68310-576CS 425.99 USD InStock
CA94025-802 68310-576 CA68310-574 94025-802 68310-574 CA68310-580 68310-580 CA33502-126 33502-126 CA47734-200 47734-200 CA68310-576
SATWipes® Sigma™ Wipes, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
The SATWIPES® Sigma™ consists of a perforated roll of saturated wipes which are dispensed through a center-pull canister. The wipes are made of 55% cellulose and 45% polyester creating highly sorbent and durable wipe.
  • Hydraulically entangled fabric creates a soft yet durable wipe
  • Clean wipe offers consistent solvent application
  • Low levels of particles and extractable contaminants to reduce contamination possibility
The isopropanol solvent blends provide safe cleaning solutions in various concentrations. Packaged with refill packs and a reusable pop-up dispenser, eliminating the need for solvent bottles and reducing solvent handling.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty