Would you like to log in again?
Canister for SATWipes® Presaturated Wipers, Contec
Supplier: Contec
SAT-CAN120
68311-030CS 116.84 USD InStock
68311-030
Canister for SATWipes® Presaturated Wipers, Contec
Wipe Dispensers
This reusable canister holds pre-saturated wipes that are 6" x 9" (152×229 mm) and are dispensed from center-pull rolls.
Canister comes with a snap close lid that helps keep wipes saturated and clean.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty