Would you like to log in again?
TAPE CR VINYL BROWN 1INX36YDS
Supplier: MICRONOVA MFG INC
CR100 PC Vinyl Tape Brown 1Inx36Yds^ Cleanroom Processed. The CR100 PC Cleanroom tape is a vinyl tape sepically processed for applications in and around the cleanroom. The tape is extremely conformable which makes it ideal for taping and sealing curved and irregular surfaces.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TAPE CR VINYL BROWN 1INX36YDS 68399-184 CR100PC-1BR Case of 36 (36 yd) Retrieving
Each(36yd) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty