Would you like to log in again?
Hand care products, Micronova
Supplier: Micronova
IC-220 IC-210 IC-110 IC-420
68400-440CS 373.6 USD InStock
68400-440 68400-374 68400-031 68400-029
Hand care products, Micronova
Skincare Soaps and Skin Cleansers
The Micronova Hand Care Products include a range of soaps and sanitizers that are formulated with highly effective, non-irritating ingredients that are ideal for frequent use in clean and controlled environments.
  • Saddle pump bottles can be used as standalone dispensers
  • Saddle pump bottles can also be used with VWR® MicroDispenser™ Touch-Free Dispenser to ensure uncontaminated results
NovaClenz™ Instant Hand Sanitizer is made with 62% Ethanol to help prevent viral transfer. Effective against bacteria, including pseudomonas, salmonella, and staphylococcus. Effective in reducing nosocomial infections. Fortified with emollients to moisturize hands.
Hy-G-Clenz™ Antimicrobial Hand Soap is made with 0.13% Benzalkonium Chloride and natural emollients. Effective against a wide range of both gram-positive and gram-negative bacteria. Persistent activity up to three hours. The soap is pH balanced and contains natural moisturizers, ideal for frequent hand washing. Meets OSHA standards.
AquaHol™ Sterile Isopropyl Alcohol Solution is made with 70% sterile IPA in 30% Wifi quality water, and meets USP requirements. Terminally filtered to 0.1 microns. Gamma Irradiated. Ideal for use over cleanroom gloves.
NovaDyne™ General-Purpose Hand Soap is a pH balanced, gentle lotion hand soap. Made with built-in moisturizers/skin emollients, it is ideal for frequent hand washing.
Certifications: Compliant with OSHA standards.
Ordering information: Pump bottles can be used as stand-alone dispensers or with the VWR® MicroDispenser touch-free soap dispenser.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty