Would you like to log in again?
ADAPTER MEGA TIP SS/DELRIN
Supplier: MICRONOVA MFG INC
MegaTip* Swab Adapter^ 10.2x3.8cm probe which, when used with a universal handle (see 68400-302, -304, -164, -166), will easily reach to clean lighting recesses, corners and angles between walls and ceilings, and other areas where a mop or wiping method is not ideal. Total length: 22.9cm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER MEGA TIP SS/DELRIN 68400-148 QDMT-1 Case of 6 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty