Would you like to log in again?
Curtain Cleaner™ Tool and Covers, Micronova
Supplier: Micronova
Curtain Cleaner™
CCSC-612 CCH-48 CCSC-6 CCSC-6IR CCSC-2
68400-178CS 674.17 USD InStock
68400-178 43300-958 68400-176 89133-452 68400-172
Curtain Cleaner™ Tool and Covers, Micronova
Mops
A spring loaded, clamp mop that works to clean and sanitize vinyl strip curtains by clamping at the top and sliding down, effectively cleaning both sides at once. Ideal for critical environments, cleanroom support, and industrial applications.
  • Cover pairs fit over two, 30.5 cm (12") bar clamps
  • Can be used with strong disinfectants
  • Launderable for re-use and autoclavable
Covers available in a choice of fabrics for specialized cleaning:
NovaPoly™ - smooth knit polyester provide excellent disinfectant dispersal.
PolySorb™ - textured polyester for removing residues and dispersing disinfectant.
NovaLite™ - latex bonded with rayon for greater friction resistance and absorbency.
The CurtainCleaner™ tool provides a simple, effective method to clean and sanitize cleanroom strip curtains and softwall enclosures, without the need for a ladder or stool. The durable stainless steel assembly includes an attached, 48" (120 cm), stainless steel handle with precise clamp hand control.
Ordering information: Mop assembly and slipcovers must be ordered separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty