Would you like to log in again?
Magid® CutMaster® Medium Weight Spectra®/Fiberglass Blend Gloves, Magid
Supplier: Magid
SP1036G-10 SP1036G-7 SP1036G-9
69100-531EA 26 USD InStock
69100-531 69100-537 69100-535
Magid® CutMaster® Medium Weight Spectra®/Fiberglass Blend Gloves, Magid
Gloves
These medium weight work gloves are manufactured from a yarn blended with composite fiberglass and Spectra, one of the highest performance fibers.
  • Low lint construction minimizes product contamination
  • Dark coloration hides dirt and extends service life
  • Reversible design allows for longer wear
  • Gloves can be laundered to increase wear time
These medium weight gloves offers protection and are reversible for longer wear. Ideal glove for work in industries.
A durable and lightweight polyethylene fiber produced using a gel-spinning process, Spectra is the cutting edge in cut-resistance, and is up to 40% stronger than other aramid fibers.
Certifications: ANSI Level A7.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty