Would you like to log in again?
VWR® Amber Latex Rubber Tubing
Supplier: VWR
VWR®
1606 1604 604R 804R 406R 808R 1012 606R 1208 1206 1004R 812 1204 2408 1006R 806R 1008 1608 402R
62996-145PK 361.9 USD InStock
62996-145 62996-178 62996-473 62996-440 62996-350 62996-382 69400-000 62996-509 62995-723 62996-418 62996-666 62996-655 62996-622 62996-699 62996-688 62996-203 62996-531 62996-564 62996-597
VWR® Amber Latex Rubber Tubing
Tubing Transfer Tubing
High-quality rubber tubing recommended for all kinds of glass connections where highly elastic, clinging tubing is desired.
  • Durometer hardness: Shore A, 35
  • Tensile strength: 3500 psi
  • Translucent and durable
Amber latex rubber tubing is available in 50' lengths on reels or in boxes.
Ordering information: This product comes in three pieces.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty