Would you like to log in again?
SatPax® 1000, Presaturated Wipers, Berkshire
Supplier: Berkshire
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SatPax®
SPX1000.001.12 SPX1000.011.12 SPX100000212 SPXC10000212 SPXC1000.00112 SPXC1000112R SPXC1000212R SPX100000112
69999-306CS 503.97 USD InStock
69999-306 76239-244 76498-148 69999-302 21912-102 21912-104 69999-300 21914-710
SatPax® 1000, Presaturated Wipers, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
SatPax® wipers provide a convenient combination of solvent and wiper that improves the cleaning process.It resolves safety issues associated with storage, mixing, and dispensing of solvents.
  • Low extractables and fiber and particle counts
  • Smooth and durable with good wet strength
  • Reduces alcohol usage and preparation/handling costs
  • Increases cleaning efficiency
SatPax® 1000 nonwoven polyester/cellulose wipers are saturated with semiconductor grade isopropyl alcohol and deionized water. Knitted polyester wipers are cleanroom laundered and are presaturated with a blend of semiconductor grade isopropyl alcohol and reverse osmosis deionized water filtered to 0.1 µm.
Certifications: ISO Class 5 – 8. Cleanroom Class 100 – 100 000. EU Grade B/C/D. USP 797.
Ordering information: Wipers are folded and packaged in a resealable cleanroom compatible pouch (containing C-folded wipers) and canisters (containing a roll of perforated wipers).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty