Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 36.5% ACS

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger
Synonyms: Muriatic acidHydrochloric acid solution

H1035-200LTB
700000-560EA 1555.5 USD InStock
700000-560
Hydrochloric acid 36.5% ACS
Hydrochloric acid
Hydrochloric Acid, Reagent, ACS, is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids, it is the least likely to be subject to an oxidation-reduction reaction. The fact that it is available as a pure reagent make it an excellent acidifying reagent. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis. As an ACS grade Reagent, Spectrum Chemical manufactured Hydrochloric Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.200
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay 36.5% Min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty