Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide solution 50% (w/w) ∼12.5 N in aqueous solution, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

S1310-1LTPL S1310-100MLPL S1310-500MLP S1310-200LTB S1310-4LTPL S1310-20LTPL
700000-752EA 2102 USD InStock
700000-752 700000-754 700000-756 700000-748 700000-758 700000-750
Sodium hydroxide solution 50% (w/w) ∼12.5 N in aqueous solution, Reagent Grade
Sodium hydroxide
Sodium Hydroxide, 50 Percent (w/w), Solution, Reagent is commonly known as Caustic soda. This inorganic compound is water soluble, as well as in ethanol and methanol. The dissolution of solid Sodium Hydroxide in water creates a highly exothermic reaction. The concentrated solution is suitable for further dilution or as a raw material feedstock in chemical synthetic transformations. As a Reagent grade item, Spectrum Chemical manufactured Sodium Hydroxide met the toughest regulatory standards for quality and purity


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.515
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty