Synonyms:
Aluminum chloride hexahydrate
AL225-100GM
AL225-50KGBL
AL225-500GM
AL225-2.5KG
AL225-12KG
95031-726EA
968
USD
InStock
95031-726
700001-084
95031-728
700001-082
95031-724
Aluminium chloride hexahydrate ≥95% USP
Aluminium chloride hexahydrate
Aluminum Chloride, Hexahydrate, USP is used for its antistatic characteristics in some excipient applications.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.