Synonyms:
Cobalt dichloride hexahydrate
, Cobalt Chloride, 6-Hydrate
, Cobaltous chloride hexahydrate
, Cobalt Chloride, Hexahydrate
C1305-2.5KG
C1305-125GM
C1305-12KG
C1305-500GM
C1305-25GM
700001-100EA
6266
USD
InStock
700001-100
700001-098
700001-108
700001-104
700001-102
Cobalt(II) chloride hexahydrate ≥98%, crystals ACS
Cobalt (II) chloride hexahydrate
Cobalt Chloride, Hexahydrate, Crystal, Reagent, ACS is an inorganic compound that has a deep purple color as a hexahydrate. It is most widely used as an indicator of water in desiccants but also has uses in organic synthesis and electroplating. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness