S1243-45KGBL
S1243-100GM
S1243-2.5KG
S1243-500GM
700001-332EA
318
USD
InStock
700001-332
700001-330
700001-328
700001-326
Sodium chlorite, anhydrous, flakes
Sodium chlorite
Sodium Chlorite, Anhydrous, Flakes is a chemical compound often used in the manufacture of paper. Used to generate chlorine dioxide for the bleaching and striping of textiles it is also frequently used as a oxidation reagent in organic synthesis.