Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl ether, anhydrous ≥99% ACS

Supplier: SPECTRUM CHEMICAL MFG CORP

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Ether2,2'-OxybisethaneEthoxyethaneEthyl Ether2,2'-OxydiethaneEthyl ether

E1010-20LT E1010-500ML E1010-1LT E1010-4LTMT
700003-390EA 972 USD InStock
700003-390 700003-392 700003-386 700003-388
Diethyl ether, anhydrous ≥99% ACS
Diethyl ether
Ethyl Ether, Anhydrous, Reagent, ACS, is also referred to as sulfuric ether, diethyl ether or even just simply as ether. This liquid was historically used as an anesthetic, but in more recent years been replaced by other more effective anesthetics. It has however some other uses such as fuel and solvent. It's an organic compound that is extremely flammable and volatile on top of being colorless. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness


Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.713
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg
Order Now
Specification Test Results
Assay 99% Min.

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty