Synonyms:
Acetic acid ammonium salt
A1120-12KG
A1120-125GM
A1120-500GM
A1120-2.5KG
700004-306EA
527
USD
InStock
700004-306
700004-310
700004-304
700004-302
Ammonium acetate ≥97%, crystals ACS
Ammonium acetate
Ammonium Acetate, Crystal, Reagent, ACS is a chemical compound that comes as a crystalline white solid. Its uses vary from biodegradable de-icer as well as being used in dialysis as part of a protein purification stage that is useful in removing contaminant via diffusion. It is also used in an aqueous solution as a buffer for ESI mass spectrometry of proteins. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness