Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lauric acid ≥97%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning
Synonyms: Dodecylic acidDodecanoic acid

L1024-500GM L1024-100GM L1024-2.5KG
700005-220EA 69 USD InStock
700005-220 700005-222 700005-224
Lauric acid ≥97%, Reagent Grade
Lauric acid
Lauric Acid, Reagent, also known as dodecanoic acid, is a medium chain fatty acid that has a vague smell of soap and is a powder. It is found naturally in human breast milk as well as cow's and goat's milk. It is generally used to produce cosmetic products but is also used in the laboratory to obtain the molar mass of substances. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.


Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 42…45 °C
Density: 0,875 g/cm³ (20 °C)
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400
Order Now
Specification Test Results
Assay 97% Min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty