Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Caffeic acid ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 3,4-Dihydroxycinnamic acid3-(3,4-Dihydroxyphenyl)acrylic acid

CA112-25KGBL CA112-25GM CA112-5GM
700005-646EA 15669 USD InStock
700005-646 700005-648 700005-644
Caffeic acid ≥99%
Caffeic acid
Caffeic Acid, also known as trans-Caffeate, is a key intermediate in the biosynthesis of lignin and is also used as a matrix in MALDI mass spectrometry.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₉H₈O₄
MW: 180.16 g/mol
Boiling Pt: 366 °C (1013 hPa)
Melting Pt: 234…237 °C
Storage Temperature: Ambient
MDL Number: MFCD00004392
CAS Number: 331-39-5
EINECS: 206-361-2
Merck Index: 12,01673
Order Now
Specification Test Results
Assay 99% Min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty