Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lactic acid racemate USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 2-Hydroxypropionic acid2-Hydroxypropanoic acidDL-Lactic acid

L1010-4LTPL-CS L1010-20LTPL L1010-4LTPL L1010-125ML L1010-20LTPL L1010-500CS6 L1010-500MLP L1010-200LTB L1010-4CS4 L1010-4LTGL
95031-442CS 2880.48 USD InStock
95031-442 700006-076 95031-514 75811-462 CA700006-076 700006-074 75811-464 95031-510 95031-444 95031-512
Lactic acid racemate USP
Lactic acid
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Density: 1.209
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Assay (C₃H₆O₃) 88.0 - 92.0%
Specific Rotation -0.05° to +0.05
Residue On Ignition max. 0.05%
Sugars To Pass Test
Chloride To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty