Synonyms:
2-Oxopropionic acid sodium salt
, α-Ketopropionic acid sodium salt
, 2-Ketopropionic acid sodium salt
, alpha-Ketopropionic acid sodium salt
, Pyruvic acid sodium salt
, Sodium 2-oxopropanoate
SO193-500GM
SO193-25KGBL
SO193-25GM
SO193-100GM
700006-598EA
935
USD
InStock
700006-598
700006-596
700006-594
700006-592
Sodium pyruvate ≥99%, Reagent Grade
Sodium pyruvate
Sodium Pyruvate, Reagent is most often added to the cell culture media as an extra source of energy. Sodium pyruvate also has a protective action against hydrogen peroxide. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.