Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish) ACS

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning
Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

E1295-25GM E1295-5GM
700010-032EA 97.92 USD InStock
700010-032 700010-030
Eosin Y (yellowish) ACS
Eosin Y (yellowish)
Eosin Y, Reagent, ACS, a form of eosin, is used as an acidic red stain for indicating cytoplasm material.

As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Melting Pt: 295…296 °C
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
EINECS: 241-409-6
Merck Index: 13,03634
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty