Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tergitol™ 15-S-9, Surfactant, Spectrum chemical

Supplier: Spectrum

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Tergitol™
T1261-500ML T1261-100ML T1261-4LTGL T1261-20LT
700011-118EA 126 USD InStock
700011-118 700011-114 700011-116 700011-110
Tergitol™ 15-S-9, Surfactant, Spectrum chemical
Cleaning Agents Detergents and Surface Cleansers
Tergitol™ 15-S-9, Surfactant is a nonionic Secondary Alcohol Ethoxylate surfactant and is an excellent emulsifier & detergent, enhances paint thickener efficiency and aids in rinse-ability of solvent-based systems. This product is suggested for use in cleaners, prewash spotters and paints & coatings.

  • Superior detergency
  • Soluble in water
  • Rapid dissolution & good rinseability
  • Low odor
  • Narrow gel range
  • Excellent formulation & handling properties
  • Readily biodegradable (OECD 301E)

Density at 20°C (68°F): 1.006 g/ml

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty