Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tergitol™ NP-7, Surfactant, Spectrum Chemical

Supplier: Spectrum

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Tergitol™
T1282-500MLG T1282-20LT T1282-100ML T1282-4LTGL
700011-148EA 72 USD InStock
700011-148 700011-154 700011-150 700011-152
Tergitol™ NP-7, Surfactant, Spectrum Chemical
Cleaning Agents Detergents and Surface Cleansers
Tergitol™ NP-7, Surfactant is a Nonylphenol Ethoxylate surfactant. It is nonionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odor. This product is suggested for use in cleaners & degreasers, prewash spotters and oil field chemicals.

  • Excellent detergency
  • Outstanding wetting
  • Versatile solubility characteristics
  • Low odor
  • Soluble in water

Density at 20°C (68°F): 1.044 g/ml

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty