Would you like to log in again?
Catalase
Supplier: Spectrum
C1328-1GM C1328-5GM
700011-380EA 733 USD InStock
700011-380 700011-376
Catalase
Enzymes
Catalase, Lyophilized, a tetramer of four polypeptide chains, is used in the food industry to remove hydrogen peroxide from milk in order to make cheese. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty