Would you like to log in again?
Papain
Supplier: Spectrum
PA125-500GM PA125-100GM
700011-474EA 200 USD InStock
700011-474 700011-476
Papain
Enzymes
Papain, Powder, Purified, is a cysteine protease enzyme the is present in the papaya fruit.
It is traditionnally used to break down meat fibers. It has more recently been used to dissociate cells in the early steps of cell culture preparation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty