Would you like to log in again?
Subtilisin A, Spectrum
Supplier: Spectrum
S1211-50ML S1211-250ML S1211-10ML
700011-490EA 795 USD InStock
700011-490 700011-488 700011-492
Subtilisin A, Spectrum
Enzymes
Subtilisin A, Aqueous Solution, a protein-digesting enzyme, can be obtained from Bacillus subtilis and is used in synthetic organic chemistry for research; it can also be found in dishwashing detergents and some cosmetics
Subtilisin A, aqueous solution, a protein-digesting enzyme, can be obtained from bacillus subtilis and is used in synthetic organic chemistry for research; it can also be found in dishwashing detergents and some cosmetics. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a certificate of analysis available.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty