Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Colophony (Rosin gum), lumps

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Rosin, gumGum rosinRosin

R1025-12KG R1025-500GM R1025-2.5KG
700011-558EA 766 USD InStock
700011-558 700011-564 700011-560
Colophony (Rosin gum), lumps
Colophony (Rosin gum)
Rosin gum in lump format, natural resin.

Rosin gum, lump is a solid derived from certain plants but mostly pines and conifers. It can be semi transparent and varies in color from black to yellow. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a certificate of analysis available.


MDL Number: MFCD00132205
CAS Number: 8050-09-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty