Would you like to log in again?
Tergitol™ TMN-6, Surfactant, Spectrum Chemical
Supplier: Spectrum
Tergitol™
T1303-100ML T1303-200LTB
700012-618EA 100000 USD InStock
700012-618 700012-616
Tergitol™ TMN-6, Surfactant, Spectrum Chemical
Cleaning Agents Detergents and Surface Cleansers
Tergitol™ TMN-6, Surfactant is a is a water-soluble, nonionic surfactant that is a superior wetting agent for use in multiple applications including paper & textile processing and hard surface cleaners.
  • 90% solution
  • Superior wetting agent
  • Penetrant & dispersant
  • Narrow gel range
  • Silicone emulsifier
Density at 20°C (68°F): 0.994 g/ml
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty