Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dextran sulfate sodium salt, Powder

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Dextran sulfate sodium from DextranDextran sulphate sodium from Dextran

DE136-100GM DE136-25GM DE136-10KGBL
700013-168EA 978 USD InStock
700013-168 700013-170 700013-174
Dextran sulfate sodium salt, Powder
Dextran sulphate sodium salt
Dextran Sulfate Sodium Salt, Powder is a sodium salt of the sulfuric acid ester of Dextran with an average molecular weight of 40,000 - 50,000. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers. It appears as a white micro-crystalline powder that is soluble in cold water.


Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Ambient
MDL Number: MFCD00081551
CAS Number: 9011-18-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty